Order Entry
Italy
Orders LinkContactUsLinkComponent
Stirring rods

Stirring rods

Stirring rods, glass, with fused ends, Glass, 6 mm, 200 mm
Catalog # 441-9586
Supplier:   DWK Life Sciences
Stirring rods
Catalog # 441-9586
Supplier:  DWK Life Sciences
Stirring rods

Stirring rods, glass, with fused ends, Glass, 6 mm, 200 mm

Price
10,80 €
Log in for availability
1 * 10 pezzi
Product Details & Documents

AR-Glas® with fused ends.

Documents

Specifications
  • Ø:
    6 mm
  • Description:
    Stirring rods, glass, with fused ends
  • Length:
    200 mm
  • Material:
    Glass