0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Italy
Orders Link
ContactUsLinkComponent
Stirrers
Stirring Rods
Stirring rods
Stirring rods
Stirring rods, glass, with fused ends, Glass, 6 mm, 150 mm
441-9585
:
DWK Life Sciences
Stirring rods
441-9585
:
DWK Life Sciences
Stirring rods
Stirring rods, glass, with fused ends, Glass, 6 mm, 150 mm
10,00 €
1 * 10 pezzi
-
+
AR-Glas® with fused ends.
SCERL143000700915_TDS.pdf
Ø:
6 mm
Description:
Stirring rods, glass, with fused ends
Length:
150 mm
Material:
Glass
Seq:
03