Instrument dishes
Lid for dishes 216-1461 and 216-1462, 360×260×20 mm
Catalog # 216-1686
Supplier: P.S. KUNSTSTOFWERKE
Jump to:
Product Details & Documents
Melamine dishes, white; PS lid, clear.
- Stable, good break-resistance
- Temperature resistant from −178 to +80 °C (melamine) and −40 to +55 °C (PS)
- Smooth surfaces for easy cleaning
Multipurpose dishes for a wide variety of laboratory applications. Also suitable for use in the food industry.
Specifications
- Description:Lid for dishes 216-1461 and 216-1462
- Seq:13
- W×D×H:360×260×20 mm



