Order Entry
Italy
Orders LinkContactUsLinkComponent
Racks for test tubes

Racks for test tubes

11 mm, 12 (2×6), 135×42×35 mm
Catalog # 212-6305
Racks for test tubes
Catalog # 212-6305
Racks for test tubes
Racks for test tubes

11 mm, 12 (2×6), 135×42×35 mm

Price
136,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Rack for test tubes, made from clear acrylic, constructed in two tiers.

  • Good thermal resistance
  • Rugged stable design
  • Easy to clean
Specifications
  • Hole Ø:
    11 mm
  • No. of Holes:
    12 (2×6)
  • W×D×H:
    135×42×35 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.