Order Entry
Italy
Orders LinkContactUsLinkComponent
Extension Cables

Extension Cables

PVC extension cable, White, 3 m
Catalog # 139-1300
Supplier:   BACHMANN ELEKTRO
Extension Cables
Catalog # 139-1300
Supplier:  BACHMANN ELEKTRO
Extension Cables
Extension Cables

PVC extension cable, White, 3 m

Price
9,94 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

PVC extension cord with moulded, double-earthed plug and Schuko coupling.

  • Cable (HO5VV-F G31,50 mm²) available in different lengths
  • Colour: white
  • Max. power: 10/16 A, 250 V~

Certifications: VDE compliant

Specifications
  • Colour:
    White
  • Description:
    PVC extension cable
  • Length:
    3 m
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.