Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Wipes for general applications, Polytex®

75 g/m², Blue, 420×420 mm, Pack of 420
Wipes for general applications, Polytex®
  115-2704
Wipes for general applications, Polytex®
Wipes for general applications, Polytex®

75 g/m², Blue, 420×420 mm, Pack of 420

 
75,50 €
   
1 * 420 pezzi
 

Soft cloths for wet wiping of sensitive surfaces.

  • 100% PP, reusable
  • Solvent resistant
  • Anti-static and fluff-free for universal lab applications and food areas
  • High tear resistance and good absorption capacity
 
  • Packed:
    Pack of 420
  • Colour:
    Blue
  • Description:
    75 g/m²
  • L×W:
    420×420 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.