Order Entry
Italy
Orders LinkContactUsLinkComponent
Wipes for general applications, Polytex®

Wipes for general applications, Polytex®

60 g/m², White, 420×420 mm, Pack of 420
Catalog # 115-2702
Wipes for general applications, Polytex®
Catalog # 115-2702
Wipes for general applications, Polytex®
Wipes for general applications, Polytex®

60 g/m², White, 420×420 mm, Pack of 420

Price
70,00 €
Log in for availability
1 * 420 pezzi
Product Details & Documents

Soft cloths for wet wiping of sensitive surfaces.

  • 100% PP, reusable
  • Solvent resistant
  • Anti-static and fluff-free for universal lab applications and food areas
  • High tear resistance and good absorption capacity
Specifications
  • Packed:
    Pack of 420
  • Colour:
    White
  • Description:
    60 g/m²
  • L×W:
    420×420 mm
Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..