Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Wipes for general applications, Polytex®

60 g/m², White, 420×420 mm, Pack of 35
Wipes for general applications, Polytex®
  115-2701
Wipes for general applications, Polytex®
Wipes for general applications, Polytex®

60 g/m², White, 420×420 mm, Pack of 35

 
5,90 €
   
1 * 35 pezzi
 

Soft cloths for wet wiping of sensitive surfaces.

  • 100% PP, reusable
  • Solvent resistant
  • Anti-static and fluff-free for universal lab applications and food areas
  • High tear resistance and good absorption capacity
 
  • Packed:
    Pack of 35
  • Colour:
    White
  • Description:
    60 g/m²
  • L×W:
    420×420 mm