Order Entry
Italy
Orders LinkContactUsLinkComponent
Safety spectacles, Op-Tema®
Safety spectacles, Op-Tema®
Anti-fog, Blue, Clear PC
Catalog # 111-1229
Supplier:   Honeywell Safety
Safety spectacles, Op-Tema®
Catalog # 111-1229
Supplier:  Honeywell Safety
Safety spectacles, Op-Tema®
Safety spectacles, Op-Tema®
Anti-fog, Blue, Clear PC
Price
10,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Comfortable safety spectacles with lightweight full view lens and built-in moulded browbar.
  • Soft, adjustable nose piece
  • Fully adjustable
  • Adjustable, break-away Flexicord retainer for added safety
Weight: 28 g
Certifications: EN 166, EN 170; 1F
Specifications
  • Type:
    Anti-fog
  • Colour:
    Blue
  • Seq:
    07
  • Lenses:
    Clear PC
Consigli
I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".
Altrimenti riceverai suggerimenti generici..