Microplates for fluorescence applications
Product Details & Documents
Designed specifically for fluorescence applications, this range of products have binding surfaces that include Microfluor® and universal binding (UB) and are available with either black or white pigmentation depending on the specific assay requirements.
- Available in 96-well or strip plate format for matching to the requirements of automated instruments
- Microfluor® microplates have raised wells to avoid cross-contamination
Black microplates have reduced background signal to improve assay sensitivity with minimised crosstalk, while avoiding light piping between the wells. They are available in high and medium binding configurations:
Microfluor® 1 Black and Black UB microplates offer low background.
Microfluor® 2 Black are irradiated microplates that provide higher binding and minimal background fluorescence.
White microplates have higher reflectivity to enhance signal and thus sensitivity, while improving signal-to-noise ratios, thus increasing the dynamic range and minimising crosstalk. They are available in high and medium binding configurations:
Microfluor® 1 White microplates offer high signal reflectance and reduced background fluorescence.
Microfluor® 2 White is an irradiated microplate for assays that require higher binding and reduced background for fluorescence assays in the ultraviolet range.
White microplates have higher reflectivity to enhance signal and thus sensitivity, while improving signal-to-noise ratios, thus increasing the dynamic range and minimising crosstalk. They are available in high and medium binding configurations:
Microfluor® 1 White microplates offer high signal reflectance and reduced background fluorescence.
Microfluor® 2 White is an irradiated microplate for assays that require higher binding and reduced background for fluorescence assays in the ultraviolet range.
Specifications
- No. of wells:96
- Colour:Black
- Description:Microfluor® 1 black plate, flat bottom wells
- Seq:03
- Cat. No.:735-0527
- Environmental certification:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #AvailabilityPrice
- Microplate lid, sterile, individually wrapped (type A)Microplate lid, sterile, individually wrapped (type A)735-0488Log in for availability1 * 50 pezzi - 253,00 €