Order Entry
Italy
Orders LinkContactUsLinkComponent
IgM purification kit , Pierce™
IgM purification kit , Pierce™
Pierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samples, 1 KIT
Catalog # PIER44897
IgM purification kit , Pierce™
Catalog # PIER44897
CAS Number:  
IgM purification kit , Pierce™
IgM purification kit , Pierce™
Pierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samples, 1 KIT
CAS Number:  
Price
1 160,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
The Pierce™ IgM Purification Kit is an easy to use affinity matrix and buffer system to purify IgM from mouse ascites fluid. The method uses immobilised mannan binding protein (MBP), which binds to IgM in the kit-supplied buffers.
  • Immobilised MBP allows specific purification of mouse IgM from ascites fluid
  • Non denaturing, near-neutral buffer conditions ensure recovery of functional antibodies
  • Stable ligand and resins can be regenerated and reused at least ten times
Delivery information: Kit includes MBP Agarose column, 5 m, 1 column; IgM Binding buffer, 800 ml; IgM Elution buffer, 500 ml; MBP Column Preparation buffer, 50 ml; and Column extender.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Pierce™ IgM Purification kit, sufficient for 10 IgM purifications, each from 0,5 ml mouse ascites samples
Specifications