Order Entry
Italy
Orders LinkContactUsLinkComponent
Substrate buffers, Pierce™
Pierce™ Stable Peroxide Substrate Buffer, 10X, 100 mL
Catalog # PIER34062
Substrate buffers, Pierce™
Catalog # PIER34062
CAS Number:  
Substrate buffers, Pierce™
Substrate buffers, Pierce™
Pierce™ Stable Peroxide Substrate Buffer, 10X, 100 mL
CAS Number:  
Price
96,50 €
Log in for availability
1 * 100 mL
Product Details & Documents
These stable peroxide and diethanolamine substrate buffers produce optimised, reliable and convenient stock solutions for HRP and alkaline phosphatase assays detection respectively.
  • Ready to use
  • Minimises assay-to-assay variability
Specifications
Specifications
  • Pk:
    100 mL
  • Description:
    Pierce™ Stable Peroxide Substrate Buffer, 10X
Specifications