Evaporating flasks
500 ml, - NS, EL-flange, uncoated
Catalog # 531-1973
Supplier: BUCHI
Jump to:
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Specifications
- Socket size:- NS
- Type:EL-flange, uncoated
- Capacity:500 ml
- Seq:31
- Cat. No.:531-1973