Order Entry
Italy
Orders LinkContactUsLinkComponent
Packaging tape, tesa®

Packaging tape, tesa®

tesa® 4204, 200,0 mm
Catalog # 819-1212
Packaging tape, tesa®
Catalog # 819-1212
Packaging tape, tesa®

tesa® 4204, 200,0 mm

Price
59,10 €
Log in for availability
1 * 1 Roll
Product Details & Documents

This tape is based on a stable PVC backing coated with a natural rubber adhesive and can be used on all common desk dispensers. Available in red, blue, green and yellow.

  • Ideal for sealing small boxes and cans
  • Can be used at bakery and flower shop counters
  • Suitable for marking based on the variety of colours available

Length: 66,0 m

Colour: Transparent

Specifications
  • Width:
    200,0 mm
  • Description:
    tesa® 4204
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.