Jump to:
Product Details & Documents
Used to stain blood, bone marrow smears, and clinical cytological specimens
The staining method according to Wright is one of the standard techniques in hematological diagnostic procedures. Like Leishman's eosin methylene blue solution and May-Grünwald's eosin methylene blue solution, it is used for staining of blood and bone marrow smears and clinical cytological specimens. The Wright’s eosin methylene blue solution is an IVD / CE product and 500 ml are sufficient for staining up to 1600 slides in Hellendahl cells.
Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
- CAS Number:68988-92-1
- MDL Number:MFCD00082143
- Density:0.80 g/cm3 (20 °C)
- Flash Pt:9.7 °C
- UN:1230
- ADR:3 (6.1),II
Documents
Merck The Color of Tissue Diagnostics
Product Literature
msds_101383
SDS
Specifications
Specification Test Results
- Suitability for microscopy (Blood smear):passes test
- Erythrocytes:pink to brownish
- Nuclei:blue-violet
- Eosinophilic granules:red to red-brown
- Neutrophilic granules:light violet
- Lymphocyte cytoplasm:Blue
Specifications
- Pack type:Plastic bottle
- Pk:1 L



