Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L

Starch Indicator Solution 1%, Standard Volumetric Solution, 6
  88632.LS
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L
  88632.LS
 :  
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L
Starch indicator solution 1%, acculute standard volumetric solution, final volume 1 L

Starch Indicator Solution 1%, Standard Volumetric Solution, 6

 :  
 
423,00 €
   
1 * 6 pezzi
 

CAS No.: 9005-84-9
Physical form: Liquid, for iodometric titrations
MDL No.: MFCD00082026

 

 

  • Pk:
    6
  • Description:
    Starch Indicator Solution 1%, Standard Volumetric Solution
  • Cat. No.:
    88632.LS
  • Physical form:
    Liquid, For iodometric titrations
  • MDL No.:
    MFCD00082026
  • CAS No.:
    9005-84-9

 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.