Order Entry
Italy
Orders LinkContactUsLinkComponent
Funnel with short stem

Funnel with short stem

35 mm, 6 mm, 35 mm
Catalog # 221-1721
Supplier:   Kavalier
Funnel with short stem
Catalog # 221-1721
Supplier:  Kavalier
Funnel with short stem
Funnel with short stem

35 mm, 6 mm, 35 mm

Price
6,70 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Borosilicate glass, grade 3.3.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With smooth internal walls.

 : DIN 12445 and ISO 4798

Specifications
  • Funnel O-Ø:
    35 mm
  • Seq:
    02
  • Stem O-Ø:
    6 mm
  • Stem length:
    35 mm