Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Stoppers, LDPE

Natural, 9,5 mm, 11 mm
  217-9248
 :   SEMADENI
Stoppers, LDPE
  217-9248
 :  SEMADENI
Stoppers, LDPE
Stoppers, LDPE

Natural, 9,5 mm, 11 mm

 
15,60 €
   
1 * 100 pezzi
 

Lamellar stoppers made from soft PE-LD.

  • Practical grip edge
  • Bright tolerance through soft lamellars

 

 
  • Ø bottom:
    9,5 mm
  • Ø top:
    11 mm
  • Colour:
    Natural
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.