Order Entry
Italy
Orders LinkContactUsLinkComponent
Stoppers, LDPE

Stoppers, LDPE

Natural, 15,5 mm, 17 mm
Catalog # 217-9246
Supplier:   SEMADENI
Stoppers, LDPE
Catalog # 217-9246
Supplier:  SEMADENI
Stoppers, LDPE
Stoppers, LDPE

Natural, 15,5 mm, 17 mm

Price
14,40 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

Lamellar stoppers made from soft PE-LD.

  • Practical grip edge
  • Bright tolerance through soft lamellars

Documents

Specifications
  • Ø bottom:
    15,5 mm
  • Ø top:
    17 mm
  • Colour:
    Natural
  • Seq:
    09