Stoppers, LDPE
Natural, 20,5 mm, 22 mm
Catalog # 217-9244
Supplier: SEMADENI
Jump to:
Product Details & Documents
Lamellar stoppers made from soft PE-LD.
- Practical grip edge
- Bright tolerance through soft lamellars
Certificates
Search by Lot NumberSpecifications
- Ø bottom:20,5 mm
- Ø top:22 mm
- Colour:Natural
- Seq:13
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



