Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Stoppers, LDPE

Natural, 8,5 mm, 10 mm
Catalog # 217-9243
Supplier:   SEMADENI
Stoppers, LDPE
Catalog # 217-9243
Supplier:  SEMADENI
Stoppers, LDPE
Stoppers, LDPE

Natural, 8,5 mm, 10 mm

Price
18,30 €
Log in for availability
1 * 100 pezzi
Product Details & Documents

Lamellar stoppers made from soft PE-LD.

  • Practical grip edge
  • Bright tolerance through soft lamellars

 

Specifications
  • Ø bottom:
    8,5 mm
  • Ø top:
    10 mm
  • Colour:
    Natural
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.