Order Entry
Italy
Orders LinkContactUsLinkComponent
Laboratory trays
Laboratory trays
225×175×50 mm, 190×140×50 mm
Catalog # 216-1361
Supplier:   BOCHEM
Laboratory trays
Catalog # 216-1361
Supplier:  BOCHEM
Laboratory trays
Laboratory trays
225×175×50 mm, 190×140×50 mm
Price
121,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Stainless steel, ¹⁸/₁₀.
  • Rounded corners for fast, easy cleaning
  • Polished electrolytically, anti-magnetic
  • Good chemical resistance, excellent thermal resistance
Multipurpose trays for a wide range of uses. Ideal as developing trays.
Specifications
  • W×D×H ext.:
    225×175×50 mm
  • Seq:
    01
  • W×D×H int.:
    190×140×50 mm