Lithium hexafluorosilicate
Glass bottle for solids, 25 g
Supplier: Thermo Fisher Scientific
Lithium hexafluorosilicate
Catalog # 39332.14
Supplier: Thermo Fisher Scientific
CAS Number: 17347-95-4
Jump to:
Product Details & Documents
CAS No.: 17347-95-4 Molecular Formula: Li2SiF6 Formula Weight: 155.96 Physical Form: Powder Odour: Odourless UN No.: 2856 MDL No.: MFCD00049473
- CAS Number:17347-95-4
- Formula:F₆Li₂Si
- MDL Number:MFCD00049473
- Molecular Weight:155.96 g/mol
- Storage Temperature:Ambient
- EINECS:241-372-6
- UN:2856
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



