Lithium hexafluorosilicate
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Lithium hexafluorosilicate
Catalog # 39332.06
Supplier: Thermo Fisher Scientific
CAS Number: 17347-95-4
Jump to:
Product Details & Documents
CAS No.: 17347-95-4 Molecular Formula: Li2SiF6 Formula Weight: 155.96 Physical Form: Powder Odour: Odourless UN No.: 2856 MDL No.: MFCD00049473
- CAS Number:17347-95-4
- Formula:F₆Li₂Si
- MDL Number:MFCD00049473
- Molecular Weight:155.96 g/mol
- Storage Temperature:Ambient
- EINECS:241-372-6
- UN:2856
- ADR:6.1,III
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



