Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Infrared optical windows

Crystal optic disc, polished both sides, Potassium bromide, 13 mm, 1 mm
  38445.KT
Infrared optical windows
  38445.KT
Infrared optical windows
Infrared optical windows

Crystal optic disc, polished both sides, Potassium bromide, 13 mm, 1 mm

 
92,20 €
   
1 * 1 pezzi
 

These optical crystal windows are ideal for infrared spectroscopy.

  • Disc or rectangular
  • Polished or unpolished

A polishing kit is available for those who wish to polish their windows.

 
  • Window material:
    Potassium bromide
  • Description:
    Crystal optic disc, polished both sides
  • Dimensions:
    13 mm
  • Seq:
    62
  • Thickness:
    1 mm
Accessories

 

  •  
    Description
     
     
  • 39798.KT
    Crystal polishing kit
       
    1 * 1 pezzi - 626,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.