Lecithin, refined
Product Details & Documents
Lecithin is used as an emulsifying and stabilizing agent in the food, pharmaceutical, and cosmetic industries. Lecithin is taken as a medicine and is also used in the manufacturing of medicines. Soybean lecithin is useful for its application in liquid ram semen. Fractionation process of sunflower lecithin with absolute ethanol was used for obtaining enriched fractions in certain phospholipids.
CAS No.: 8002-43-5 Physical Form: Powder Odour: Odourless Beilstein Reference: 5209585 Merck Reference: 14,5428 MDL No.: MFCD00147406
Caution: RUO – Research Use Only
- CAS Number:8002-43-5
- MDL Number:MFCD00147406
- Merck Index:14,5428
- Density:1.0305 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:232-307-2
Documents
Specifications
Specification Test Results
- Appearance (Color):Light tan to medium yellow
- Form:Powder
- Assay (unspecified):Acetone insoluble ≥ 97%
- Acid Value:<35 mg KOH/g
- Residual water:< 1%
- Peroxide content:5 meq /kg, max
Specifications
- Pack type:Plastic bottle
- Pk:250 g
Specifications
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



