Order Entry
Italy
Orders LinkContactUsLinkComponent
Acetone ≥99.5% semiconductor grade

Acetone ≥99.5% semiconductor grade

Glass bottle, 4x1 L, 4
Catalog # 19392.M1
Flammable
Irritant
Acetone ≥99.5% semiconductor grade
Catalog # 19392.M1
CAS Number:  67-64-1
Acetone ≥99.5% semiconductor grade
Acetone ≥99.5% semiconductor grade

Glass bottle, 4x1 L, 4

CAS Number:  67-64-1
Price
153,00 €
Log in for availability
1 * 4 pezzi
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Purity: 99.5% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 63580 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765

Synonyms:  2-Propanone, DMK, Propanone, Dimethyl ketone
  • CAS Number:67-64-1
  • Formula:CH₃COCH₃
  • MDL Number:MFCD00008765
  • Molecular Weight:58.08 g/mol
  • Merck Index:14,66
  • Boiling Pt:56 °C
  • Melting Pt:–94 °C
  • Density:0.791
  • Flash Pt:–17 °C (1 °F)
  • Storage Temperature:Ambient
  • EINECS:200-662-2
  • UN:1090
  • ADR:3,II
  • REACH:01-2119471330-49
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    4
  • Pk Info:
    4x1 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.