Order Entry
Italy
Orders LinkContactUsLinkComponent
Barium chromate ≥98%
Glass bottle for solids, 250 g
  14669.30
Barium chromate ≥98%
  14669.30
 :  10294-40-3
Barium chromate ≥98%
Barium chromate ≥98%
Glass bottle for solids, 250 g
 :  10294-40-3
76,30 €
1 * 250 g
Purity: 98% CAS No.: 10294-40-3 Molecular Formula: BaCrO4 Formula Weight: 253.33 Physical Form: Powder Density: 4.50 Appearance: Yellow Odour: Odourless Solubility: Insoluble in water, acetic acid, chromic acid Merck Reference: 14,972 UN No.: 1479 MDL No.: MFCD00014180
  •  :10294-40-3
  •  :BaCrO₄
  •  :MFCD00014180
  •  :253.32 g/mol
  •  :14,972
  •  :1400 °C
  •  :4.50
  •  :Ambient
  •  :233-660-5
  •  :1479
  •  :5.1,III
  •  :Density: 4,5 g/cm³ (25 °C)
  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g