Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium peroxide ≥95%

Lithium peroxide ≥95%

Glass bottle for solids, 100 g
Catalog # 13421.22
Oxidizing
Corrosive
Lithium peroxide ≥95%
Catalog # 13421.22
CAS Number:  12031-80-0
Lithium peroxide ≥95%
Lithium peroxide ≥95%

Glass bottle for solids, 100 g

CAS Number:  12031-80-0
Price
468,00 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 95% CAS No.: 12031-80-0 Molecular Formula: Li2O2 Formula Weight: 45.88 Physical Form: Powder Sensitivity: Air and Moisture Sensitive Appearance: White to off-white Odour: Odourless UN No.: 1472 MDL No.: MFCD00016184

  • CAS Number:12031-80-0
  • Formula:Li₂O₂
  • MDL Number:MFCD00016184
  • Molecular Weight:45.88 g/mol
  • Density:1.2063 g/cm³ (18 °C)
  • Storage Temperature:Ambient
  • EINECS:234-758-0
  • UN:1472
  • ADR:5.1,II
  •  :Density: 1.2063 g/cm³ (18 °C)
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.