Order Entry
Italy
Orders LinkContactUsLinkComponent
Lithium carbonate ≥99%

Lithium carbonate ≥99%

Glass bottle for solids, 10 kg
Codice catalogo 13418.A9
Irritant
Nome del fornitore:   Thermo Fisher Scientific
Lithium carbonate ≥99%
Codice catalogo 13418.A9
Nome del fornitore:  Thermo Fisher Scientific
Numero CAS:  554-13-2
Lithium carbonate ≥99%
Lithium carbonate ≥99%

Glass bottle for solids, 10 kg

Numero CAS:  554-13-2
Price
2 290,00 €
Log in for availability
1 * 10 kg
Product Details & Documents

Purity: 99% CAS No.: 554-13-2 Molecular Formula: Li2CO3 Formula Weight: 73.90 Physical Form: Powder Density: 2.11 Appearance: White Odour: Odourless pH: 10-11.5 Melting Point: 723° Boiling Point: 1310°dec. Solubility: Soluble in water and dilute acids. Insoluble in alcohol Merck Reference: 14,5527 MDL No.: MFCD00011084

  • CAS Number:554-13-2
  • Formula:Li₂CO₃
  • MDL Number:MFCD00011084
  • Molecular Weight:73.89 g/mol
  • Merck Index:14,5527
  • Boiling Pt:1310 °C (dec.)
  • Melting Pt:723 °C
  • Density:2.11
  • Storage Temperature:Ambient
  • EINECS:209-062-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.