Selenium dioxide ≥99.4% (metals basis)
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Selenium dioxide ≥99.4% (metals basis)
Catalog # 12358.36
Supplier: Thermo Fisher Scientific
CAS Number: 7446-08-4
Jump to:
Product Details & Documents
Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odour: Odourless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562
As an oxidizing agent, in manufacturing of other selenium compounds
- CAS Number:7446-08-4
- Formula:SeO₂
- MDL Number:MFCD00003562
- Molecular Weight:110.96 g/mol
- Merck Index:14,8434
- Melting Pt:340…350 °C (subl.)
- Density:3.95
- Storage Temperature:Ambient
- EINECS:231-194-7
- UN:3283
- ADR:6.1,II
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



