Order Entry
Italy
Orders LinkContactUsLinkComponent
Selenium dioxide ≥99.4% (metals basis)

Selenium dioxide ≥99.4% (metals basis)

Glass bottle for solids, 100 g
Catalog # 12358.22
Toxic
Health
Environment
Selenium dioxide ≥99.4% (metals basis)
Catalog # 12358.22
CAS Number:  7446-08-4
Selenium dioxide ≥99.4% (metals basis)
Selenium dioxide ≥99.4% (metals basis)

Glass bottle for solids, 100 g

CAS Number:  7446-08-4
Price
64,60 €
Log in for availability
1 * 100 g
Product Details & Documents

Purity: 99.4% CAS No.: 7446-08-4 Molecular Formula: O2Se Formula Weight: 110.96 Physical Form: Powder Density: 3.95 Appearance: White to pink Odour: Odourless Melting Point: 340-350° subl. Solubility: Partially soluble in water, methanol, ethanol, acetone. Soluble in concentrated H2SO4 Merck Reference: 14,8434 UN No.: 3283 MDL No.: MFCD00003562

As an oxidizing agent, in manufacturing of other selenium compounds

  • CAS Number:7446-08-4
  • Formula:SeO₂
  • MDL Number:MFCD00003562
  • Molecular Weight:110.96 g/mol
  • Merck Index:14,8434
  • Melting Pt:340…350 °C (subl.)
  • Density:3.95
  • Storage Temperature:Ambient
  • EINECS:231-194-7
  • UN:3283
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.