1-Hexene for synthesis
Glass bottle, 2,5 L
: Merck
- :592-41-6
- :C₆H₁₂
- :MFCD00009505
- :84.16 g/mol
- :63 °C (1013 hPa)
- :–140 °C
- :0.675 g/cm³ (20 °C)
- :–26 °C
- :Ambient
- :209-753-1
- :2370
- :3,II
- Pack type:Glass bottle
- Pk:2,5 L