Order Entry
Italy
Orders LinkContactUsLinkComponent
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)

Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)

Plastic drum, 25 L
Catalog # 9713.9025
Health
Irritant
Supplier:   VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Catalog # 9713.9025
Supplier:  VWR Chemicals
CAS Number:  50-00-0
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)

Plastic drum, 25 L

CAS Number:  50-00-0
Price
292,00 €
Log in for availability
1 * 25 L
Product Details & Documents

(= 10% Formaline solution)

Synonyms:  Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  •  :Stabilised with methanol 0,5-1,5 %
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specification Test Results

  • Assay (Formaldehyde):
    3.5 - 4.5 %
  • pH (20 °C):
    6.9 - 7.1
  • Methanol:
    0.5 - 1.5 %

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    25 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.