Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Plastic jerrican, 5 L
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7.0 ±0.1)
Catalog # 9713.5000
Supplier: VWR Chemicals
CAS Number: 50-00-0
Jump to:
Product Details & Documents
(= 10% Formaline solution)
Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde
- CAS Number:50-00-0
- Formula:CH₂O
- MDL Number:MFCD00003274
- Molecular Weight:30.03 g/mol
- :Stabilised with methanol 0,5-1,5 %
- Storage Temperature:Ambient
- EINECS:200-001-8
Documents
Certificates
Search by Lot Number10L Plastic drum for liquids DIN 51 UN (ZPACK13951000) 1L Plastic bottle for liquids DIN 45 UN X (ZPACK13961000) 25L Plastic drum for liquids DIN 61 (ZPACK13952500)
Packaging Information
Specifications
Specification Test Results
- Assay (Formaldehyde):3.5 - 4.5 %
- pH (20 °C):6.9 - 7.1
- Methanol:0.5 - 1.5 %
Specifications
- Pack type:Plastic jerrican
- Pk:5 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



