Order Entry
Italy
Orders LinkContactUsLinkComponent
High purity dNTPs

High purity dNTPs

2'-Deoxycytidine 5'-Triphosphate, sodium (dCTP), 250 mg
Catalog # 27-1860-04
Supplier:   Cytiva
High purity dNTPs
Catalog # 27-1860-04
Supplier:  Cytiva
CAS Number:  
High purity dNTPs
High purity dNTPs

2'-Deoxycytidine 5'-Triphosphate, sodium (dCTP), 250 mg

CAS Number:  
Price
587,00 €
Log in for availability
1 * 250 mg
Product Details & Documents

dNTPs designed for applications requiring high purity such as amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

  • High purity dNTPs are functionally tested in long PCR to be PCR-qualified, and meet or exceed criteria for high-quality sequencing with thermo sequenase DNA polymerase
  • High purity (>98% triphosphate) confirmed by chromatographic analysis and ³¹P nuclear magnetic resonance (NMR) spectroscopy

Ordering information: Supplied as solids. For high purity solutions dNTPs, see illustra™ dNTP solutions.

Specifications

Specifications

  • Pk:
    250 mg
  • Description:
    2'-Deoxycytidine 5'-Triphosphate, sodium (dCTP)
  • Cat. No.:
    27-1860-04
  • Storage conditions:
    -20°C
  • Purity:
    ≥ 98% triphosphate

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.