Order Entry
Italy
Orders LinkContactUsLinkComponent
Holders for dissecting needles

Holders for dissecting needles

Holder for dissecting needles, with plastic handle, 22 g, 230 mm
Catalog # 391-0099
Supplier:   USBECK Laborgeräte
Holders for dissecting needles
Catalog # 391-0099
Holders for dissecting needles
Holders for dissecting needles

Holder for dissecting needles, with plastic handle, 22 g, 230 mm

Price
9,60 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Needle holders made from nickel plated brass. Either with or without plastic handle.

Specifications
  • Description:
    Holder for dissecting needles, with plastic handle, 22 g
  • Length:
    230 mm
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.