Order Entry
Italy
Orders LinkContactUsLinkComponent
Clamps with chain
Clamps with chain
40 - 120 mm
Catalog # 241-2016
Supplier:   USBECK Laborgeräte
Clamps with chain
Catalog # 241-2016
Clamps with chain
Clamps with chain
40 - 120 mm
Price
45,90 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Zinc die-cast, nickel plated. For attaching glass beakers, flasks, reaction vessels and other laboratory vessels to support stands.
  • With cork-lined bracket
  • Adjustable, rubber covered chain
  • Without support stand bosshead
Specifications
  • Spanning width:
    40 - 120 mm