Order Entry
Italy
Orders LinkContactUsLinkComponent
Clamps
Clamps
30 - 80 mm
Catalog # 241-0254
Supplier:   USBECK Laborgeräte
Clamps
Catalog # 241-0254
Clamps
Clamps
30 - 80 mm
Price
19,20 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Zinc die-cast, powder coated.
  • The jaws of the clamps are covered with layer of cork to protect the clamped items
  • Without bosshead for fixing to stand
  • With wing screws for rapid easy clamping
Specifications
  • Spanning width:
    30 - 80 mm