Order Entry
Italy
Orders LinkContactUsLinkComponent
Burner, Teclu

Burner, Teclu

Teclu burner with air regulation needle valve, natural gas, head Ø 32 mm, tubing Ø 19 mm, Natural gas, 32 mm
Catalog # 451-1366
Supplier:   USBECK Laborgeräte
Burner, Teclu
Catalog # 451-1366
Burner, Teclu
Burner, Teclu

Teclu burner with air regulation needle valve, natural gas, head Ø 32 mm, tubing Ø 19 mm, Natural gas, 32 mm

Price
101,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Nickel plated brass base, in powder coated grey.

  • With air regulation valve and throttle screw
  • Without pilot light

Height: 165 mm

Specifications
  • Description:
    Teclu burner with air regulation needle valve, natural gas, head Ø 32 mm, tubing Ø 19 mm
  • Head Ø:
    32 mm
  • For:
    Natural gas
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.