Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Hexene for synthesis

1-Hexene for synthesis

Glass bottle, 1 L
Catalog # 8.20639.1000
Flammable
Health
Supplier:   Merck
1-Hexene for synthesis
Catalog # 8.20639.1000
Supplier:  Merck
CAS Number:  592-41-6
1-Hexene for synthesis

Glass bottle, 1 L

CAS Number:  592-41-6
Price
77,40 €
Log in for availability
1 * 1 L
Product Details & Documents
  • CAS Number:592-41-6
  • Formula:C₆H₁₂
  • MDL Number:MFCD00009505
  • Molecular Weight:84.16 g/mol
  • Boiling Pt:63 °C (1013 hPa)
  • Melting Pt:–140 °C
  • Density:0.675 g/cm³ (20 °C)
  • Flash Pt:–26 °C
  • Storage Temperature:Ambient
  • EINECS:209-753-1
  • UN:2370
  • ADR:3,II

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.