Order Entry
Italy
Orders LinkContactUsLinkComponent
Comparator kits, multiparameter for water testing, Comparator 2000+

Comparator kits, multiparameter for water testing, Comparator 2000+

Drinking water, AF 357, Chloride: 0 - 5000 mg/l Cl
Chlorine: 0,02 - 0,3 mg/l Cl₂
Chlorine: 0,2 - 4 mg/l Cl₂
Fluoride: 0 - 1,6 mg/l F
Hardness: 0 - 500 mg/l CaCO₃
Hazen colour: 10 - 90 mg Pt/l
pH value: 6 - 8,4
Catalog # 705-0787
Supplier:   Lovibond
Comparator kits, multiparameter for water testing, Comparator 2000+
Catalog # 705-0787
Supplier:  Lovibond
Comparator kits, multiparameter for water testing, Comparator 2000+
Comparator kits, multiparameter for water testing, Comparator 2000+

Drinking water, AF 357, Chloride: 0 - 5000 mg/l Cl
Chlorine: 0,02 - 0,3 mg/l Cl₂
Chlorine: 0,2 - 4 mg/l Cl₂
Fluoride: 0 - 1,6 mg/l F
Hardness: 0 - 500 mg/l CaCO₃
Hazen colour: 10 - 90 mg Pt/l
pH value: 6 - 8,4

Price
2 610,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents

Lovibond® Comparator system 2000+ is an extremely versatile, modular system for testing water. The battery operated portable lighting unit can be integrated to give standardised lighting conditions to enhance reproducibility.

  • Compact and portable
  • Simple to use yet uncompromising in terms of precision and reproducibility of results

 : Comparator test kits are supplied as a complete system in a sturdy plastic case. Together with the Comparator 2000+ and test discs, each kit includes all the necessary cells, accessories and Lovibond® tablet reagents (for 100 measurements). The AF368 is not supplied with reagents.

Specifications
  • Range:
    Chloride: 0 - 5000 mg/l Cl
    Chlorine: 0,02 - 0,3 mg/l Cl₂
    Chlorine: 0,2 - 4 mg/l Cl₂
    Fluoride: 0 - 1,6 mg/l F
    Hardness: 0 - 500 mg/l CaCO₃
    Hazen colour: 10 - 90 mg Pt/l
    pH value: 6 - 8,4
  • Description:
    Drinking water
  • Seq:
    02
  • Type:
    AF 357