Order Entry
Italy
Orders LinkContactUsLinkComponent
Precision probes, T

Precision probes, T

Stainless steel probe (IP 67) with PTFE cable, −50…+350 °C, ±0,2 (−2…+70);
±1,0 °C, 4×125 mm
Catalog # 628-3295
Supplier:   TESTO
Precision probes, T
Catalog # 628-3295
Supplier:  TESTO
Precision probes, T
Precision probes, T

Stainless steel probe (IP 67) with PTFE cable, −50…+350 °C, ±0,2 (−2…+70);
±1,0 °C, 4×125 mm

Price
135,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Specifications
  • Range:
    −50…+350 °C
  • Description:
    Stainless steel probe (IP 67) with PTFE cable
  • Seq:
    05
  • Accuracy:
    ±0,2 (−2…+70);
    ±1,0 °C
  • Ø×L:
    4×125 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.