Active Mmp-2 Enzyme 1 * 5 µG
Catalog # PF023-5
Supplier: Merck Millipore (Oncogene)
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
Tissue DNA kit, genomic DNA isolation from tissue, E.Z.N.A.®
Supplier: OMEGA BIO-TEK
VWR®, Sampling Spoons
Supplier: VWR Collection
Decontamination solution, DNA AWAY®
Supplier: Thermo Fisher Scientific
VWR® Standard Line Transfer Pipettes
Supplier: VWR Collection
GelRed® & GelGreen® Nucleic acid gel stains
Supplier: Biotium
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Petri Dish, PS
Supplier: VWR Collection
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Supplier: EPPENDORF
VWR®, Straight Sample Containers, with Screw Cap
Supplier: VWR Collection
VWR® PCR Tubes, 0,2 ml
Supplier: VWR Collection
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
Anti-IL-12A Rabbit Monoclonal Antibody [clone: EPR5736]
Supplier: Abcam