Order Entry
Italy
Orders LinkContactUsLinkComponent
Modules, Nunc-Immuno™ LockWell™
Modules, Nunc-Immuno™ LockWell™
MaxiSorp™, U8×12, Clear, 96
Catalog # 735-0027
   Low Manufacturing Impact
Modules, Nunc-Immuno™ LockWell™
Catalog # 735-0027
   Low Manufacturing Impact
Modules, Nunc-Immuno™ LockWell™
Modules, Nunc-Immuno™ LockWell™
MaxiSorp™, U8×12, Clear, 96
Price
650,00 €
Log in for availability
1 * 60 pezzi
Product Details & Documents
PS. Nunc-Immuno™ LockWell™ modules facilitate assays from large scale screening to single-well tests. Available with a choice of MaxiSorp™, PolySorp™, MediSorp™ and MultiSorp™ surfaces. All MaxiSorp™ modules are certified for binding homogeneity. Framed modules (external dimensions: 128×86 mm) fit all standard equipment.
  • Each well locks in the frame at the same height to prevent jamming in automated systems
  • Strips are easy to take out or put in the frame, without use of any special tools
MaxiSorp™ surface has a high affinity to molecules with mixed hydrophilic/hydrophobic domains; PolySorp™ surface has a high affinity for hydrophobic molecules; MediSorp™ has a surface chemistry intermediate between PolySorp™ and MaxiSorp™, providing low background signal with samples containing serum; MultiSorp™ surfaces have a high affinity to hydrophilic molecules.
Colour coded modules available on request, please visit website or contact your local Avantor sales office for further information.
Specifications
  • No. of wells:
    96
  • Colour:
    Clear
  • Description:
    MaxiSorp™, U8×12
  • Seq:
    06
  • Cat. No.:
    735-0027
  • Environmental certification:
    ISO 14001 (Certificate Registration No.CERT-IT000330)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • LockWell™ frame
    LockWell™ frame
    732-2689
    Log in for availability
    1 * 60 pezzi - 111,00 €