Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
Competent cells, expression strains, B834
Competent cells, expression strains, B834
B834(DE3)pLysS competent Cells, 5×200 μl
Catalog # 69042-4
Competent cells, expression strains, B834
Catalog # 69042-4
Competent cells, expression strains, B834
B834(DE3)pLysS competent Cells, 5×200 μl
Price
314,00 €
Log in for availability
1 * 1 mL
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A common method for transformation of plasmids into E.coli is the use of chemically competent cells. Although competent cells can be prepared in the laboratory, greater efficiency, reproducibility, and convenience are achieved using Novagen® prepared competent cells. Every Novagen® competent cell strain is verified for phenotype and purity, and is guaranteed for transformation efficiency.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilising pET recombinants encoding target proteins that affect cell growth and viability.
The standard volume per tube is 0,2 ml, which provides enough cells for 10 transformations. All kits are supplied with SOC medium and test plasmid. Several strains are also available as Singles™ Competent Cells, which are provided in single-use, 50 μl volumes for extra convenience and efficiency.
B834 is a methionine auxotroph that allows high specific activity labeling of target proteins with ³⁵S-methionine or selenomethionine for crystallography studies.
Specifications
  • Descrizione:
    B834(DE3)pLysS competent Cells, 5×200 μl
  • Seq:
    04