Fmoc-D-Pro-OH
Plastic bottle for solids, 25 g
Catalog # 04-13-1031-25
Supplier: Novabiochem (Part of Merck)
Jump to:
Product Details & Documents
Synonyms: N-Fmoc-D-proline
- CAS Number:101555-62-8
- Formula:C₂₀H₁₉NO₄
- MDL Number:MFCD00077067
- Molecular Weight:337.38 g/mol
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



