Preparative sorbents, LiChroprep®
LiChroprep® RP-18, 15 - 25 µm, 02, 500 g
Supplier: MERCK PRODUCTION CHEMICALS
Preparative sorbents, LiChroprep®
Catalog # 1.13901.0500
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Jump to:
Product Details & Documents
LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.
- Reproducible homogeneous silica gel matrix
- Narrow, well defined particle size distribution for high performance and high permeability
- Excellent selectivity and efficiency
- Large number of applications
- Large batch size
- Comprehensive regulatory documents are available
LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.
Specifications
Specifications
- Particle Size:15 - 25 µm
- Pk:500 g
- Pore size:02
- Description:LiChroprep® RP-18



