Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Preparative sorbents, LiChroprep®

LiChroprep® RP-18, 40 - 63 µm, 08, 250 g
Catalog # 1.13900.0250
Preparative sorbents, LiChroprep®
Catalog # 1.13900.0250
CAS Number:  
Preparative sorbents, LiChroprep®

LiChroprep® RP-18, 40 - 63 µm, 08, 250 g

CAS Number:  
Price
1 800,00 €
Log in for availability
1 * 250 g
Product Details & Documents

LiChroprep® is a proven, highly successful packing material providing fast, effective and reproducible separations.

  • Reproducible homogeneous silica gel matrix
  • Narrow, well defined particle size distribution for high performance and high permeability
  • Excellent selectivity and efficiency
  • Large number of applications
  • Large batch size
  • Comprehensive regulatory documents are available

LiChroprep® is one of the most successful and reliable sorbents used in HPLC and medium pressure chromatography.

Specifications

Specifications

  • Particle Size:
    40 - 63 µm
  • Pk:
    250 g
  • Pore size:
    08
  • Description:
    LiChroprep® RP-18

Specifications