Order Entry
Italy
Orders LinkContactUsLinkComponent
Bioindicator, Sterikon® plus
Bioindicator, Sterikon® plus
Sterikon® plus bioindicator
Catalog # 1.10274.0001
Supplier:   Merck
Bioindicator, Sterikon® plus
Catalog # 1.10274.0001
Supplier:  Merck
Bioindicator, Sterikon® plus
Bioindicator, Sterikon® plus
Sterikon® plus bioindicator
Price
86,00 €
Log in for availability
1 * 15 Ampoll
Product Details & Documents
Sterikon® plus bioindicators can be used to test the functionality of autoclaves (15 minutes at 121 °C). They can also be used to check any sort of autoclave load for the success of sterilisation. For example, pharmaceuticals, particularly products in ampoules, canned food and more.
  • Easy interpretation due to colour change
  • Safe, because spores of non-pathogenic organisms (Geobacillus stearothermophilus) are used
Acc. to the recommendation of USP and EP for the use of bioindicators for checking the autoclaving of pharmaceutical products.
Certifications: According to the recommendation of USP and EP for the use of bioindicators for checking the autoclaving of pharmaceutical products.
Specifications
  • Description:
    Sterikon® plus bioindicator